Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMK1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020397.3 | NP_065130.1 | 2 | 388 | ||
NM_153498.3 | NP_705718.1 | 2 | 388 | |||
XM_006717481.3 | XP_006717544.1 | 2 | 652 | |||
XM_006717482.3 | XP_006717545.1 | 2 | 417 | |||
XM_006717483.3 | XP_006717546.1 | 2 | 379 | |||
XM_011519591.2 | XP_011517893.1 | 2 | 825 | |||
XM_017016438.1 | XP_016871927.1 | 3 | 683 | |||
XM_017016439.1 | XP_016871928.1 | 4 | 514 | |||
GenBank mRNA | AB081726.1 | - | 2 | 344 | ||
AF286366.1 | - | 2 | 199 | |||
AK289393.1 | - | 3 | 199 | |||
BC035745.1 | - | 2 | 205 | 5748490 | ||
BU542765.1 | - | 2 | 449 | |||
HY168807.1 | - | 2 | 389 |