Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020404.2 | NP_065137.1 | 1 | 1765 | ||
GenBank mRNA | AF279142.1 | - | 1 | 1753 | ||
AJ295846.1 | - | 1 | 1765 | |||
AK027290.1 | - | 1 | 887 | |||
BC051340.1 | - | 1 | 1752 | 6339237 | ||
BC104484.1 | - | 1 | 1765 | 40007855 | ||
BC105633.1 | - | 1 | 1765 | 40007854 | ||
KU215671.1 | - | 1 | 1703 | |||
KU257670.1 | - | 1 | 1697 |