Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020405.4 | NP_065138.2 | 5 | 793 | ||
GenBank mRNA | AF279144.2 | - | 5 | 663 | ||
AF378753.1 | - | 5 | 663 | |||
AK093589.1 | - | 5 | 814 | |||
AK127539.1 | - | 4 | 610 | |||
AK303699.1 | - | 4 | 480 | |||
AK312999.1 | - | 5 | 615 | |||
AY704670.1 | - | 5 | 663 | |||
AY704671.1 | - | 5 | 663 | |||
AY704672.1 | - | 5 | 609 | |||
BC036059.1 | - | 5 | 759 | 5298499 | ||
KU257674.1 | - | 5 | 521 |