Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RTN4 SIRNA Assays ›
Gene Symbol:
RTN4
Gene Name:
reticulon 4
Gene Aliases:
ASY, NI220/250, NOGO, NOGO-A, NOGOC, NSP, NSP-CL, Nbla00271, Nbla10545, Nogo-B, Nogo-C, RTN-X, RTN4-A, RTN4-B1, RTN4-B2, RTN4-C
Chromosome Location:
Chr.2: 54972187 - 55137831 on Build GRCh38
UniGene ID:
Hs.637850
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s32768
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RTN4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321859.1
NP_001308788.1
4
2705
NM_001321860.1
NP_001308789.1
4
2775
NM_001321861.1
NP_001308790.1
4
2738
NM_001321862.1
NP_001308791.1
4
2725
NM_001321863.1
NP_001308792.1
4
2675
NM_001321904.1
NP_001308833.1
5
3153
NM_007008.2
NP_008939.1
2
395
NM_020532.4
NP_065393.1
4
3458
NM_153828.2
NP_722550.1
2
1001
NM_207520.1
NP_997403.1
3
1058
NM_207521.1
NP_997404.1
4
2716
NR_135829.1
2
281
NR_135830.1
2
259
XM_005264434.3
XP_005264491.1
5
2852
XM_017004518.1
XP_016860007.1
5
2891
XM_017004519.1
XP_016860008.1
5
3008
GenBank mRNA
AB015639.1
-
2
769
AB020693.1
-
4
3294
AB040462.1
-
4
3404
AB040463.1
-
2
947
AB073351.1
-
2
1130
AF063601.1
-
2
1718
AF077050.1
-
2
394
AF087901.1
-
2
286
AF125103.1
-
2
395
AF132047.1
-
2
882
AF132048.1
-
2
301
AF148537.1
-
4
3301
AF148538.1
-
2
844
AF177332.1
-
2
300
AF320999.1
-
3
2464
AF333336.1
-
4
2720
AJ251383.1
-
4
3160
AJ251384.1
-
2
703
AJ251385.1
-
2
181
AK075039.1
-
2
935
AK129806.1
-
2
860
AK130812.1
-
2
762
AY102276.1
-
2
415
AY102277.1
-
2
947
AY102278.1
-
3
1004
AY102279.1
-
4
3404
AY123245.1
-
4
2717
AY123246.1
-
4
2775
AY123247.1
-
4
2738
AY123248.1
-
4
2724
AY123249.1
-
4
2685
AY123250.1
-
4
2675
BC001035.2
-
2
366
BC007109.1
-
2
292
BC010737.1
-
2
708
BC012619.1
-
2
858
BC014366.1
-
2
292
BC016165.1
-
3
1007
BC026788.1
-
2
889
BC068991.1
-
2
951
BC071848.1
-
3
689
BC150182.1
-
4
3408
40148210
BC152425.1
-
4
3294
8862539
BC152555.1
-
4
3408
40148984
CN429712.1
-
2
230
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
axonal fasciculation
cerebral cortex radial glia guided migration
negative regulation of cell growth
negative regulation of axon extension
regulation of apoptotic process
negative regulation of axonogenesis
nuclear pore complex assembly
cardiac epithelial to mesenchymal transition
endoplasmic reticulum tubular network organization
endoplasmic reticulum tubular network assembly
cell-cell adhesion
regulation of branching morphogenesis of a nerve
protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0