Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000088.3 | NP_000079.2 | 11, 12 | 938 | ||
XM_005257058.3 | XP_005257115.2 | 11, 12 | 947 | |||
XM_005257059.4 | XP_005257116.2 | 11, 12 | 947 | |||
XM_011524341.1 | XP_011522643.1 | 11, 12 | 947 | |||
GenBank mRNA | AB209597.1 | - | 11, 12 | 913 | ||
BC036531.2 | - | 11, 12 | 928 | 5264710 | ||
JQ236861.1 | - | 11, 12 | 812 | |||
M36546.1 | - | 11, 12 | 895 | |||
X07884.1 | - | 11, 12 | 895 | |||
Z74615.1 | - | 11, 12 | 931 |