Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGAPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282917.1 | NP_001269846.1 | 3 | 417 | ||
NM_001282918.1 | NP_001269847.1 | 3 | 559 | |||
NM_020336.3 | NP_065069.1 | 3 | 559 | |||
XM_005260462.2 | XP_005260519.1 | 3 | 517 | |||
XM_005260463.2 | XP_005260520.1 | 3 | 517 | |||
XM_005260464.2 | XP_005260521.1 | 3 | 517 | |||
XM_005260465.2 | XP_005260522.1 | 3 | 517 | |||
XM_017027966.1 | XP_016883455.1 | 3 | 517 | |||
XM_017027967.1 | XP_016883456.1 | 3 | 517 | |||
XM_017027969.1 | XP_016883458.1 | 3 | 505 | |||
GenBank mRNA | AB033045.3 | - | 3 | 549 | ||
AB621815.1 | - | 2 | 339 | |||
AK123576.1 | - | 2 | 218 | |||
AK304242.1 | - | 3 | 515 | |||
BC047482.1 | - | 3 | 522 | 5270777 | ||
CR936800.1 | - | 3 | 323 | |||
DR002520.1 | - | 3 | 566 |