Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMK1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020439.2 | NP_065172.1 | 8 | 774 | ||
XM_017001866.1 | XP_016857355.1 | 8 | 917 | |||
XM_017001867.1 | XP_016857356.1 | 8 | 768 | |||
GenBank mRNA | AF428261.1 | - | 8 | 744 | ||
AK095713.1 | - | 8 | 917 | |||
AK289675.1 | - | 8 | 757 | |||
AL049688.1 | - | 7 | 689 | |||
AY212935.1 | - | 7 | 680 | |||
BC032787.1 | - | 8 | 741 | 5179957 |