Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMIZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020338.3 | NP_065071.1 | 11 | 1516 | ||
XM_005269987.4 | XP_005270044.1 | 11 | 1626 | |||
XM_005269988.3 | XP_005270045.1 | 11 | 1557 | |||
XM_006717923.3 | XP_006717986.1 | 11 | 1554 | |||
XM_006717924.3 | XP_006717987.1 | 10 | 1476 | |||
XM_006717925.3 | XP_006717988.1 | 11 | 1557 | |||
XM_011539978.2 | XP_011538280.1 | 5 | 830 | |||
XM_011539979.1 | XP_011538281.1 | 5 | 956 | |||
XM_011539980.2 | XP_011538282.1 | 5 | 784 | |||
XM_017016440.1 | XP_016871929.1 | 5 | 1776 | |||
XM_017016441.1 | XP_016871930.1 | 5 | 734 | |||
XM_017016442.1 | XP_016871931.1 | 5 | 956 | |||
XM_017016443.1 | XP_016871932.1 | 5 | 776 | |||
GenBank mRNA | AB033050.2 | - | 5 | 735 | ||
AF357909.1 | - | 5 | 662 | |||
AK024490.1 | - | 4 | 674 | |||
AY235683.1 | - | 8 | 1013 | |||
BC007288.1 | - | 11 | 1516 |