Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167882.1 | NP_001161354.1 | 2, 3 | 506 | ||
NM_020337.2 | NP_065070.1 | 3, 4 | 1781 | |||
XM_017008471.1 | XP_016863960.1 | 2, 3 | 2514 | |||
GenBank mRNA | AK127496.1 | - | 2, 3 | 632 | ||
AK291068.1 | - | 3, 4 | 1781 | |||
AK295135.1 | - | 2, 3 | 506 | |||
BX640674.1 | - | 2, 3 | 298 |