Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081550.1 | NP_001075019.1 | 35 | 4523 | ||
XM_005262450.3 | XP_005262507.1 | 35 | 4523 | |||
XM_017029662.1 | XP_016885151.1 | 37 | 4835 | |||
XM_017029663.1 | XP_016885152.1 | 37 | 4835 | |||
XM_017029664.1 | XP_016885153.1 | 37 | 4835 | |||
XM_017029665.1 | XP_016885154.1 | 37 | 4835 | |||
XM_017029666.1 | XP_016885155.1 | 37 | 4823 | |||
XM_017029667.1 | XP_016885156.1 | 37 | 4820 | |||
XM_017029668.1 | XP_016885157.1 | 37 | 4820 | |||
XM_017029669.1 | XP_016885158.1 | 36 | 4769 | |||
XM_017029670.1 | XP_016885159.1 | 36 | 4757 | |||
XM_017029671.1 | XP_016885160.1 | 36 | 4754 | |||
XM_017029672.1 | XP_016885161.1 | 37 | 4739 | |||
XM_017029673.1 | XP_016885162.1 | 36 | 4697 | |||
XM_017029674.1 | XP_016885163.1 | 36 | 4685 | |||
XM_017029675.1 | XP_016885164.1 | 36 | 4673 | |||
XM_017029676.1 | XP_016885165.1 | 36 | 4673 | |||
XM_017029677.1 | XP_016885166.1 | 35 | 4631 | |||
XM_017029678.1 | XP_016885167.1 | 35 | 4619 | |||
XM_017029679.1 | XP_016885168.1 | 35 | 4535 | |||
GenBank mRNA | AF441770.1 | - | 30 | 4146 | ||
BC072400.1 | - | 7 | 1036 | |||
BX648654.1 | - | 5 | 1230 |