Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCOLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020533.2 | NP_065394.1 | 2 | 269 | ||
GenBank mRNA | AF249319.1 | - | 2 | 212 | ||
AF287269.1 | - | 2 | 253 | |||
AJ293659.1 | - | 2 | 208 | |||
AJ293970.1 | - | 2 | 208 | |||
AK026102.1 | - | 2 | 223 | |||
AK222673.1 | - | 2 | 248 | |||
AK310109.1 | - | 2 | 244 | |||
BC005149.2 | - | 2 | 251 | 3507836 | ||
DC356874.1 | - | 2 | 272 |