Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288951.1 | NP_001275880.1 | 5 | 1067 | ||
NM_001288953.1 | NP_001275882.1 | 6 | 1058 | |||
NM_001288955.1 | NP_001275884.1 | 5 | 1067 | |||
NM_020458.3 | NP_065191.2 | 5 | 1067 | |||
XM_005264439.3 | XP_005264496.1 | 6 | 1220 | |||
XM_011532998.2 | XP_011531300.1 | 7 | 1285 | |||
XM_011532999.2 | XP_011531301.1 | 5 | 1010 | |||
XM_011533000.2 | XP_011531302.1 | 2 | 142 | |||
XM_017004524.1 | XP_016860013.1 | 5 | 1011 | |||
XM_017004525.1 | XP_016860014.1 | 5 | 552 | |||
XM_017004526.1 | XP_016860015.1 | 5 | 1011 | |||
XM_017004527.1 | XP_016860016.1 | 5 | 1162 | |||
XM_017004528.1 | XP_016860017.1 | 6 | 1227 | |||
XM_017004529.1 | XP_016860018.1 | 5 | 1010 | |||
XM_017004530.1 | XP_016860019.1 | 5 | 1011 | |||
GenBank mRNA | AB032966.2 | - | 6 | 647 | ||
AK056464.1 | - | 6 | 1030 | |||
AL834383.1 | - | 5 | 1067 | |||
BC065554.1 | - | 2 | 185 | |||
BC111487.1 | - | 5 | 955 | 6041916 | ||
BC114365.2 | - | 5 | 698 | 40038218 |