Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020665.5 | NP_065716.1 | 3 | 389 | ||
XM_017029680.1 | XP_016885169.1 | 3 | 257 | |||
XM_017029681.1 | XP_016885170.1 | 3 | 664 | |||
GenBank mRNA | AF229179.1 | - | 3 | 156 | ||
AK313835.1 | - | 3 | 214 | |||
AY359060.1 | - | 3 | 139 | |||
BC014317.1 | - | 3 | 393 | 4048217 | ||
BC015099.1 | - | 3 | 221 | 3829035 | ||
BC050606.1 | - | 3 | 141 | 5183554 |