Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020673.2 | NP_065724.1 | 4 | 509 | ||
GenBank mRNA | AF091034.2 | - | 4 | 487 | ||
AJ276210.1 | - | 4 | 474 | |||
AK091180.1 | - | 4 | 263 | |||
BC015710.1 | - | 4 | 434 | 3907891 | ||
BC063457.1 | - | 4 | 493 | 5182786 | ||
BI457862.1 | - | 4 | 514 | |||
BT007046.1 | - | 4 | 247 |