Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMRAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305141.1 | NP_001292070.1 | 3 | 507 | ||
NM_001305142.1 | NP_001292071.1 | 3 | 494 | |||
NM_020677.4 | NP_065728.1 | 3 | 608 | |||
XM_006720906.2 | XP_006720969.1 | 4 | 500 | |||
XM_011522567.2 | XP_011520869.1 | 3 | 498 | |||
XM_017023485.1 | XP_016878974.1 | 2 | 471 | |||
XM_017023486.1 | XP_016878975.1 | 2 | 474 | |||
XM_017023487.1 | XP_016878976.1 | 2 | 474 | |||
XM_017023488.1 | XP_016878977.1 | 3 | 583 | |||
XM_017023489.1 | XP_016878978.1 | 4 | 596 | |||
XM_017023491.1 | XP_016878980.1 | 2 | 474 | |||
XM_017023492.1 | XP_016878981.1 | 3 | 583 | |||
GenBank mRNA | AF225419.1 | - | 3 | 613 | ||
AK098784.1 | - | 3 | 468 | |||
BC002927.2 | - | 3 | 388 | 3954035 | ||
BC007364.2 | - | 3 | 450 | 3630659 |