Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029884.1 | NP_001025055.1 | 17 | 3754 | ||
XM_005267064.3 | XP_005267121.1 | 16 | 5833 | |||
XM_005267065.3 | XP_005267122.1 | 18 | 3780 | |||
XM_005267066.2 | XP_005267123.1 | 16 | 4637 | |||
XM_006715521.1 | XP_006715584.1 | 18 | 4007 | |||
XM_006715522.3 | XP_006715585.1 | 16 | 5874 | |||
XM_011535981.2 | XP_011534283.1 | 16 | 6630 | |||
XM_011535982.2 | XP_011534284.1 | 15 | 6591 | |||
XM_011535983.2 | XP_011534285.1 | 17 | 5055 | |||
XM_017011102.1 | XP_016866591.1 | 15 | 4600 | |||
GenBank mRNA | AB033035.1 | - | 10 | 2627 | ||
AK294798.1 | - | 16 | 3222 | |||
BC089428.1 | - | 12 | 2865 | |||
BC140864.1 | - | 15 | 3491 | 9021732 | ||
BC151134.1 | - | 15 | 3491 | 9057034 |