Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020733.1 | NP_065784.1 | 6 | 2234 | ||
XM_005247666.1 | XP_005247723.1 | 7 | 2529 | |||
GenBank mRNA | AB033063.2 | - | 5 | 1829 | ||
BC067235.1 | - | 1 | 127 |