Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLGN4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282145.1 | NP_001269074.1 | 6 | 1908 | ||
NM_001282146.1 | NP_001269075.1 | 5 | 1583 | |||
NM_020742.3 | NP_065793.1 | 5 | 1418 | |||
NM_181332.2 | NP_851849.1 | 5 | 1601 | |||
XM_005274564.2 | XP_005274621.1 | 6 | 1655 | |||
XM_005274565.2 | XP_005274622.1 | 6 | 1638 | |||
XM_005274566.4 | XP_005274623.1 | 6 | 1582 | |||
XM_006724504.3 | XP_006724567.1 | 6 | 1523 | |||
XM_011545547.2 | XP_011543849.1 | 6 | 1657 | |||
XM_011545548.2 | XP_011543850.1 | 6 | 1821 | |||
XM_017029690.1 | XP_016885179.1 | 6 | 1467 | |||
XM_017029691.1 | XP_016885180.1 | 5 | 1597 | |||
XM_017029692.1 | XP_016885181.1 | 5 | 1522 | |||
XM_017029693.1 | XP_016885182.1 | 5 | 1407 | |||
GenBank mRNA | AB033086.1 | - | 5 | 1418 | ||
AF376803.1 | - | 5 | 1418 | |||
AK022621.1 | - | 2 | 252 | |||
AK055471.1 | - | 6 | 1641 | |||
AK291036.1 | - | 5 | 1455 | |||
AX773938.1 | - | 6 | 1529 | |||
AY358562.1 | - | 5 | 1006 | |||
BC034018.1 | - | 5 | 1384 | 4702963 |