Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020754.3 | NP_065805.2 | 12 | 2981 | ||
XM_006713714.3 | XP_006713777.1 | 12 | 3005 | |||
XM_017006955.1 | XP_016862444.1 | 11 | 2920 | |||
GenBank mRNA | AB033030.1 | - | 12 | 2981 | ||
AK293726.1 | - | 12 | 2578 | |||
BC112163.1 | - | 12 | 2476 | 8327631 | ||
BC112165.1 | - | 12 | 2476 | 8327633 |