Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304840.1 | NP_001291769.1 | 22 | 3970 | ||
NM_020760.2 | NP_065811.1 | 24 | 4304 | |||
XM_006712646.3 | XP_006712709.1 | 24 | 4362 | |||
XM_006712648.3 | XP_006712711.1 | 23 | 4295 | |||
XM_017004563.1 | XP_016860052.1 | 24 | 4341 | |||
GenBank mRNA | AB037722.1 | - | 24 | 4304 | ||
AL390186.1 | - | 1 | 69 | |||
BC117194.1 | - | 24 | 4220 | 40125745 | ||
BC117198.1 | - | 24 | 4220 | 40125749 | ||
CR749424.1 | - | 22 | 4068 |