Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020780.1 | NP_065831.1 | 13 | 2801 | ||
XM_011541828.2 | XP_011540130.1 | 12 | 4903 | |||
XM_011541829.1 | XP_011540131.1 | 12 | 1776 | |||
XM_011541830.2 | XP_011540132.1 | 13 | 2884 | |||
XM_011541831.2 | XP_011540133.1 | 13 | 2884 | |||
GenBank mRNA | AB037758.1 | - | 13 | 2801 | ||
AB593117.1 | - | 13 | 2884 | |||
AL117236.1 | - | 12 | 1721 |