Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL8A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005202.3 | NP_005193.1 | 3 | 226 | ||
XM_005270477.3 | XP_005270534.1 | 3 | 926 | |||
GenBank mRNA | AK074129.1 | - | 1 | 144 | ||
BC096295.1 | - | 1 | 144 | |||
BC096296.3 | - | 1 | 144 | 40007335 |