Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282660.1 | NP_001269589.1 | 4 | 415 | ||
NM_001282661.1 | NP_001269590.1 | 7 | 827 | |||
NM_001282662.1 | NP_001269591.1 | 7 | 827 | |||
NM_001318139.1 | NP_001305068.1 | 5 | 552 | |||
NM_020831.4 | NP_065882.1 | 7 | 827 | |||
XM_005261692.1 | XP_005261749.1 | 6 | 632 | |||
XM_005261694.1 | XP_005261751.1 | 6 | 722 | |||
XM_011530283.1 | XP_011528585.1 | 8 | 935 | |||
XM_011530285.1 | XP_011528587.1 | 8 | 846 | |||
XM_017028888.1 | XP_016884377.1 | 7 | 910 | |||
XM_017028889.1 | XP_016884378.1 | 3 | 1910 | |||
GenBank mRNA | AB037859.2 | - | 7 | 799 | ||
AF368061.1 | - | 7 | 752 | |||
AJ297257.1 | - | 4 | 409 | |||
AJ297258.1 | - | 7 | 809 | |||
BC115039.1 | - | 7 | 793 | 5551767 | ||
BC139730.1 | - | 5 | 466 | |||
CR456522.1 | - | 4 | 226 | |||
HG764171.1 | - | 5 | 552 |