Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282660.1 | NP_001269589.1 | 9 | 2224 | ||
NM_001282661.1 | NP_001269590.1 | 11 | 2486 | |||
NM_001282662.1 | NP_001269591.1 | 12 | 2636 | |||
NM_001318139.1 | NP_001305068.1 | 10 | 2361 | |||
NM_020831.4 | NP_065882.1 | 12 | 2636 | |||
XM_005261692.1 | XP_005261749.1 | 11 | 2441 | |||
XM_005261694.1 | XP_005261751.1 | 11 | 2531 | |||
XM_011530283.1 | XP_011528585.1 | 13 | 2744 | |||
XM_011530285.1 | XP_011528587.1 | 13 | 2655 | |||
XM_011530287.1 | XP_011528589.1 | 5 | 1623 | |||
XM_017028888.1 | XP_016884377.1 | 12 | 2719 | |||
XM_017028889.1 | XP_016884378.1 | 8 | 3719 | |||
GenBank mRNA | AB037859.2 | - | 12 | 2608 | ||
AF368061.1 | - | 12 | 2561 | |||
AJ297257.1 | - | 9 | 2218 | |||
AJ297258.1 | - | 12 | 2618 | |||
AL713710.1 | - | 4 | 1431 | |||
BC115039.1 | - | 12 | 2602 | 5551767 | ||
CR456522.1 | - | 9 | 2035 | |||
HG764171.1 | - | 10 | 2361 |