Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOMEZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020834.2 | NP_065885.2 | 2 | 1233 | ||
GenBank mRNA | AB037864.1 | - | 2 | 1159 | ||
AF463523.1 | - | 2 | 1157 | |||
AK296927.1 | - | 2 | 1193 | |||
AK301849.1 | - | 2 | 1079 | |||
AK302788.1 | - | 2 | 1093 | |||
BC130392.1 | - | 1 | 1058 | 40146423 |