Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAHCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291324.1 | NP_001278253.1 | 25, 26 | 6322 | ||
XM_011525063.2 | XP_011523365.1 | 26, 27 | 6690 | |||
XM_017024899.1 | XP_016880388.1 | 24, 25 | 6279 | |||
GenBank mRNA | AB040880.1 | - | 21, 22 | 4073 |