Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173602.2 | NP_775873.2 | 14, 15 | 1871 | ||
XM_005269044.1 | XP_005269101.1 | 14, 15 | 1874 | |||
XM_006719520.3 | XP_006719583.1 | 15, 16 | 1847 | |||
XM_006719521.3 | XP_006719584.1 | 14, 15 | 1684 | |||
XM_011538594.2 | XP_011536896.1 | 15, 16 | 1900 | |||
XM_011538596.2 | XP_011536898.1 | 14, 15 | 1687 | |||
XM_017019699.1 | XP_016875188.1 | 15, 16 | 1882 | |||
XM_017019700.1 | XP_016875189.1 | 14, 15 | 1856 | |||
XM_017019701.1 | XP_016875190.1 | 14, 15 | 1853 | |||
XM_017019703.1 | XP_016875192.1 | 13, 14 | 1650 | |||
XM_017019704.1 | XP_016875193.1 | 13, 14 | 1658 | |||
XM_017019705.1 | XP_016875194.1 | 13, 14 | 1640 | |||
GenBank mRNA | AB040896.2 | - | 5, 6 | 485 | ||
AK126758.1 | - | 3, 4 | 242 |