Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020857.2 | NP_065908.1 | 4 | 1546 | ||
XM_011521843.2 | XP_011520145.1 | 3 | 1076 | |||
XM_011521844.2 | XP_011520146.1 | 4 | 1651 | |||
XM_017022447.1 | XP_016877936.1 | 5 | 935 | |||
GenBank mRNA | AB040908.1 | - | 4 | 1521 | ||
AF308802.1 | - | 4 | 1521 | |||
AL713725.1 | - | 4 | 1546 | |||
BC001513.2 | - | 4 | 1373 | 2822780 |