Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029939.3 | NP_001025110.2 | 6 | 1026 | ||
NM_001167583.2 | NP_001161055.1 | 5 | 867 | |||
NM_001174157.1 | NP_001167628.1 | 4 | 758 | |||
NM_001174158.1 | NP_001167629.1 | 5 | 867 | |||
NM_001289394.1 | NP_001276323.1 | 6 | 1103 | |||
NM_020863.3 | NP_065914.2 | 5 | 944 | |||
NR_110323.1 | 5 | 944 | ||||
XM_011517203.1 | XP_011515505.1 | 5 | 841 | |||
XM_011517204.2 | XP_011515506.1 | 4 | 664 | |||
XM_011517206.1 | XP_011515508.1 | 5 | 865 | |||
XM_017013716.1 | XP_016869205.1 | 4 | 739 | |||
GenBank mRNA | AB040918.1 | - | 3 | 342 | ||
AB167738.1 | - | 5 | 933 | |||
AB167739.1 | - | 6 | 1092 | |||
AB167740.1 | - | 6 | 1019 | |||
AB167741.1 | - | 5 | 860 | |||
AK094089.1 | - | 4 | 741 | |||
AK300133.1 | - | 5 | 856 | |||
AK300324.1 | - | 5 | 863 | |||
AK300471.1 | - | 5 | 929 | |||
AK311494.1 | - | 5 | 865 | |||
AL832066.1 | - | 5 | 944 | |||
BC012989.2 | - | 5 | 858 | |||
BC025423.2 | - | 3 | 536 | |||
BC098405.1 | - | 5 | 867 | |||
BC101766.1 | - | 5 | 857 | 8069272 | ||
BC101768.1 | - | 5 | 857 | 8069274 | ||
BC143519.1 | - | 4 | 672 | 9052028 |