Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPP10 SIRNA Assays ›
Gene Symbol:
DPP10
Gene Name:
dipeptidyl peptidase like 10
Gene Aliases:
DPL2, DPPY, DPRP-3, DPRP3
Chromosome Location:
Chr.2: 114442358 - 115845752 on Build GRCh38
UniGene ID:
Hs.580539
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s33475
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPP10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001004360.3
NP_001004360.2
20
1950
NM_001178034.1
NP_001171505.1
20
1877
NM_001178036.1
NP_001171507.1
21
2354
NM_001178037.1
NP_001171508.1
20
1920
NM_001321905.1
NP_001308834.1
21
2380
NM_001321906.1
NP_001308835.1
21
1996
NM_001321907.1
NP_001308836.1
19
2226
NM_001321908.1
NP_001308837.1
20
2239
NM_001321909.1
NP_001308838.1
19
1854
NM_001321910.1
NP_001308839.1
21
2323
NM_001321911.1
NP_001308840.1
20
2322
NM_001321912.1
NP_001308841.1
21
2354
NM_001321913.1
NP_001308842.1
19
2150
NM_001321914.1
NP_001308843.1
20
1875
NM_020868.4
NP_065919.2
20
2265
XM_017004565.1
XP_016860054.1
20
2385
XM_017004566.1
XP_016860055.1
19
1869
GenBank mRNA
AB040925.2
-
20
1832
AK025075.1
-
8
676
AK289967.1
-
20
1964
AK296490.1
-
20
1952
AK315147.1
-
20
2069
AY172661.3
-
20
1844
AY387785.1
-
20
1866
BC030832.1
-
20
2301
4811771
DQ857321.1
-
20
1832
DQ857322.1
-
20
1856
DQ857323.1
-
20
1823
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
protein localization to plasma membrane
positive regulation of establishment of protein localization to plasma membrane
regulation of potassium ion transmembrane transport
serine-type peptidase activity
dipeptidyl-peptidase activity
potassium channel regulator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0