Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020884.4 | NP_065935.3 | 10 | 1074 | ||
XM_006723840.3 | XP_006723903.1 | 8 | 715 | |||
XM_017027986.1 | XP_016883475.1 | 8 | 715 | |||
GenBank mRNA | AB040945.2 | - | 8 | 783 | ||
BC151242.1 | - | 8 | 795 | 8860515 |