Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142641.1 | NP_001136113.1 | 17 | 3623 | ||
XM_005266171.3 | XP_005266228.1 | 20 | 3854 | |||
XM_005266173.3 | XP_005266230.1 | 20 | 3849 | |||
XM_005266175.3 | XP_005266232.1 | 19 | 3788 | |||
XM_005266176.3 | XP_005266233.1 | 19 | 3730 | |||
XM_005266177.3 | XP_005266234.1 | 20 | 3723 | |||
XM_011534803.2 | XP_011533105.1 | 20 | 3913 | |||
XM_011534804.2 | XP_011533106.1 | 20 | 3910 | |||
XM_011534805.2 | XP_011533107.1 | 19 | 3844 | |||
XM_011534806.2 | XP_011533108.1 | 19 | 3847 | |||
XM_011534807.2 | XP_011533109.1 | 19 | 3789 | |||
XM_011534808.2 | XP_011533110.1 | 20 | 3782 | |||
XM_011534809.2 | XP_011533111.1 | 19 | 3725 | |||
XM_011534810.2 | XP_011533112.1 | 19 | 3703 | |||
XM_011534812.2 | XP_011533114.1 | 21 | 3735 | |||
GenBank mRNA | AB046765.2 | - | 17 | 3394 |