Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL17A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000494.3 | NP_000485.3 | 50 | 3778 | ||
GenBank mRNA | AB900157.1 | - | 49 | 3609 | ||
M91669.1 | - | 50 | 3714 |