Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL17A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000494.3 | NP_000485.3 | 4 | 360 | ||
GenBank mRNA | AB900157.1 | - | 3 | 191 | ||
AK309940.1 | - | 4 | 306 | |||
BC004478.2 | - | 4 | 342 | |||
M91669.1 | - | 4 | 296 |