Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020943.2 | NP_065994.1 | 15, 16 | 1842 | ||
XM_005246726.2 | XP_005246783.1 | 15, 16 | 1945 | |||
GenBank mRNA | AB046824.1 | - | 14, 15 | 1661 | ||
AK025635.1 | - | 15, 16 | 1699 | |||
AK026978.1 | - | 8, 9 | 799 | |||
AK123647.1 | - | 2, 3 | 799 | |||
BC016651.1 | - | 14, 15 | 1666 | |||
BC031216.1 | - | 15, 16 | 1842 | |||
BC041662.1 | - | 8, 9 | 793 | |||
BC053573.1 | - | 12, 13 | 1364 | |||
BC057826.1 | - | 15, 16 | 1700 | |||
BC093952.1 | - | 14, 15 | 1564 | 7939797 | ||
BC093954.1 | - | 14, 15 | 1564 | 7939799 | ||
BC143433.1 | - | 13, 14 | 1647 |