Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIER1 SIRNA Assays ›
Gene Symbol:
MIER1
Gene Name:
MIER1 transcriptional regulator
Gene Aliases:
ER1, MI-ER1
Chromosome Location:
Chr.1: 66924895 - 66988619 on Build GRCh38
UniGene ID:
Hs.605432
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s33645
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIER1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077700.2
NP_001071168.2
9
940
NM_001077701.2
NP_001071169.1
8
796
NM_001077702.2
NP_001071170.2
10
1014
NM_001077703.2
NP_001071171.2
9
940
NM_001077704.2
NP_001071172.1
8
796
NM_001146110.1
NP_001139582.1
10
1017
NM_001146111.1
NP_001139583.1
10
1014
NM_001146112.1
NP_001139584.1
10
975
NM_001146113.1
NP_001139585.1
8
794
NM_020948.3
NP_065999.2
10
1014
XM_005271076.3
XP_005271133.1
10
1052
XM_011541864.2
XP_011540166.1
11
959
XM_011541865.2
XP_011540167.1
11
920
XM_017001919.1
XP_016857408.1
11
936
XM_017001920.1
XP_016857409.1
11
953
XM_017001921.1
XP_016857410.1
11
981
XM_017001922.1
XP_016857411.1
11
956
XM_017001923.1
XP_016857412.1
9
776
XM_017001924.1
XP_016857413.1
10
908
XM_017001925.1
XP_016857414.1
10
1013
XM_017001926.1
XP_016857415.1
11
952
XM_017001927.1
XP_016857416.1
9
825
XM_017001928.1
XP_016857417.1
9
901
XM_017001929.1
XP_016857418.1
10
882
XM_017001930.1
XP_016857419.1
9
826
XM_017001931.1
XP_016857420.1
10
860
XM_017001932.1
XP_016857421.1
8
789
XM_017001933.1
XP_016857422.1
7
643
XM_017001934.1
XP_016857423.1
6
539
GenBank mRNA
AB046830.1
-
6
565
AF515446.1
-
8
796
AF515447.1
-
9
877
AF515448.2
-
10
951
AK302061.1
-
9
940
AK307717.1
-
10
911
AL831987.2
-
10
956
AY124186.2
-
10
951
AY124187.1
-
9
877
AY124188.1
-
8
796
AY124189.2
-
10
951
AY124190.1
-
9
877
AY124191.1
-
8
796
AY124192.2
-
10
951
AY124193.1
-
9
877
AY124194.1
-
8
796
BC108726.1
-
10
895
5194296
BC125217.1
-
7
592
40123797
BC125218.1
-
7
592
40123803
CR627441.1
-
10
851
Back To Top
More Information
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
signal transduction
positive regulation of chromatin silencing
positive regulation of I-kappaB kinase/NF-kappaB signaling
DNA binding
signal transducer activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0