Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL19A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001858.5 | NP_001849.2 | 6, 7 | 778 | ||
XM_011535437.2 | XP_011533739.1 | 6, 7 | 1045 | |||
XM_011535438.2 | XP_011533740.1 | 6, 7 | 773 | |||
XM_017010253.1 | XP_016865742.1 | 6, 7 | 1045 | |||
XM_017010254.1 | XP_016865743.1 | 6, 7 | 1045 | |||
XM_017010255.1 | XP_016865744.1 | 6, 7 | 984 | |||
XM_017010256.1 | XP_016865745.1 | 6, 7 | 778 | |||
XM_017010257.1 | XP_016865746.1 | 6, 7 | 1045 | |||
XM_017010258.1 | XP_016865747.1 | 6, 7 | 778 | |||
XM_017010259.1 | XP_016865748.1 | 6, 7 | 1044 | |||
GenBank mRNA | AK310621.1 | - | 6, 7 | 768 | ||
BC113362.1 | - | 6, 7 | 746 | 8322414 | ||
BC113364.1 | - | 6, 7 | 746 | 8322416 | ||
D38163.1 | - | 6, 7 | 778 | |||
L12347.1 | - | 2, 3 | 346 | |||
M63597.1 | - | 5, 6 | 587 |