Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGDCC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020962.2 | NP_066013.1 | 8 | 1666 | ||
XM_011521845.2 | XP_011520147.1 | 8 | 1846 | |||
XM_011521846.2 | XP_011520148.1 | 8 | 1470 | |||
XM_011521847.1 | XP_011520149.1 | 8 | 1530 | |||
XM_017022448.1 | XP_016877937.1 | 8 | 1848 | |||
XM_017022449.1 | XP_016877938.1 | 8 | 1854 | |||
GenBank mRNA | AB046848.1 | - | 2 | 726 | ||
AB052622.1 | - | 8 | 1666 |