Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172231.3 | NP_757386.2 | 6 | 696 | ||
XM_005260002.2 | XP_005260059.1 | 5 | 711 | |||
XM_011528155.2 | XP_011526457.1 | 5 | 589 | |||
GenBank mRNA | AF521128.1 | - | 6 | 696 | ||
AK096152.1 | - | 5 | 785 | |||
AK292085.1 | - | 6 | 693 | |||
AK294618.1 | - | 6 | 758 | |||
AY072916.1 | - | 6 | 872 | |||
AY072917.1 | - | 6 | 676 | |||
BC063784.1 | - | 6 | 674 | |||
BC142988.1 | - | 6 | 693 | 8860435 |