Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021185.4 | NP_067008.3 | 11 | 1358 | ||
XM_005259114.4 | XP_005259171.1 | 11 | 1454 | |||
XM_005259115.4 | XP_005259172.1 | 11 | 1377 | |||
XM_006723308.3 | XP_006723371.1 | 11 | 1974 | |||
XM_006723311.2 | XP_006723374.1 | 11 | 1379 | |||
XM_011527174.2 | XP_011525476.1 | 11 | 1368 | |||
XM_011527175.2 | XP_011525477.1 | 11 | 1274 | |||
XM_011527177.2 | XP_011525479.1 | 11 | 1238 | |||
XM_011527178.2 | XP_011525480.1 | 12 | 1720 | |||
XM_011527179.1 | XP_011525481.1 | 11 | 1358 | |||
XM_011527182.2 | XP_011525484.1 | 10 | 1102 | |||
XM_017027054.1 | XP_016882543.1 | 9 | 971 | |||
GenBank mRNA | AK128220.1 | - | 11 | 1141 | ||
AK310460.1 | - | 7 | 688 | |||
AL133023.1 | - | 11 | 1974 | |||
BC094785.1 | - | 11 | 1278 | |||
BC109035.1 | - | 7 | 757 | 40028284 |