Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021185.4 | NP_067008.3 | 19 | 2296 | ||
XM_005259114.4 | XP_005259171.1 | 19 | 2392 | |||
XM_005259115.4 | XP_005259172.1 | 19 | 2315 | |||
XM_005259116.2 | XP_005259173.1 | 17 | 2116 | |||
XM_006723308.3 | XP_006723371.1 | 19 | 2912 | |||
XM_006723310.2 | XP_006723373.1 | 19 | 2179 | |||
XM_006723311.2 | XP_006723374.1 | 19 | 2317 | |||
XM_011527174.2 | XP_011525476.1 | 19 | 2306 | |||
XM_011527175.2 | XP_011525477.1 | 19 | 2212 | |||
XM_011527176.2 | XP_011525478.1 | 19 | 2215 | |||
XM_011527177.2 | XP_011525479.1 | 19 | 2176 | |||
XM_011527178.2 | XP_011525480.1 | 20 | 2658 | |||
XM_011527179.1 | XP_011525481.1 | 19 | 2296 | |||
XM_011527182.2 | XP_011525484.1 | 18 | 2040 | |||
XM_017027053.1 | XP_016882542.1 | 18 | 2176 | |||
XM_017027054.1 | XP_016882543.1 | 17 | 1909 | |||
XM_017027055.1 | XP_016882544.1 | 6 | 1385 | |||
GenBank mRNA | AK128220.1 | - | 19 | 2079 | ||
AK310460.1 | - | 15 | 1626 | |||
BC094785.1 | - | 19 | 2216 | |||
BC109035.1 | - | 15 | 1695 | 40028284 |