Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021257.3 | NP_067080.1 | 2, 3 | 583 | ||
GenBank mRNA | AF422797.1 | - | 2, 3 | 583 | ||
AJ245946.1 | - | 2, 3 | 208 | |||
AK098350.1 | - | 2, 3 | 185 | |||
AK315395.1 | - | 2, 3 | 296 | |||
BC032509.1 | - | 2, 3 | 290 | 5247233 |