Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX6A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005205.3 | NP_005196.1 | 2 | 282 | ||
GenBank mRNA | BC029818.1 | - | 2 | 242 | 5174639 | |
BP319904.1 | - | 282 | ||||
F27891.1 | - | 2 | 245 | |||
HE647858.1 | - | 2 | 179 | |||
M83308.1 | - | 2 | 215 |