Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021259.2 | NP_067082.2 | 9 | 1634 | ||
GenBank mRNA | AB045292.1 | - | 9 | 1591 | ||
AK024725.1 | - | 6 | 933 | |||
AK091823.1 | - | 9 | 1412 | |||
AL546643.3 | - | 5 | 689 | |||
BC021557.1 | - | 9 | 1525 | 4899167 |