Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RXFP1 SIRNA Assays ›
Gene Symbol:
RXFP1
Gene Name:
relaxin/insulin like family peptide receptor 1
Gene Aliases:
LGR7, RXFPR1
Chromosome Location:
Chr.4: 158521711 - 158653372 on Build GRCh38
UniGene ID:
Hs.591686
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s34026
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RXFP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001253727.1
NP_001240656.1
9
1057
NM_001253728.1
NP_001240657.1
8
877
NM_001253729.1
NP_001240658.1
9
976
NM_001253730.1
NP_001240659.1
8
901
NM_001253732.1
NP_001240661.1
10
1060
NM_001253733.1
NP_001240662.1
10
1060
NM_021634.3
NP_067647.2
9
976
NR_045579.1
10
1665
NR_045580.1
9
1029
NR_045581.1
9
1000
NR_045582.1
9
1081
NR_045583.1
8
919
NR_045584.1
10
1029
XM_011532174.1
XP_011530476.1
9
1026
XM_011532175.1
XP_011530477.1
9
1026
XM_011532176.1
XP_011530478.1
9
948
XM_011532179.2
XP_011530481.1
9
1026
XM_017008517.1
XP_016864006.1
9
1026
XM_017008518.1
XP_016864007.1
9
948
XM_017008519.1
XP_016864008.1
10
990
XM_017008520.1
XP_016864009.1
9
1079
XM_017008521.1
XP_016864010.1
10
909
XM_017008522.1
XP_016864011.1
9
997
XM_017008523.1
XP_016864012.1
9
948
XM_017008524.1
XP_016864013.1
9
948
XM_017008525.1
XP_016864014.1
8
880
XM_017008526.1
XP_016864015.1
5
519
GenBank mRNA
AB208943.1
-
10
798
AF190500.1
-
9
713
AK122647.1
-
11
1017
AK294694.1
-
9
821
AK295040.1
-
10
1488
AK300379.1
-
8
772
AK307889.1
-
8
818
AK312470.1
-
10
892
AY899848.1
-
10
766
AY899849.1
-
9
713
AY899850.1
-
8
614
BC112142.1
-
9
795
8327610
BC113617.1
-
9
798
8322669
BC143556.1
-
9
818
BC143558.1
-
9
851
BC143559.1
-
8
723
BX647980.1
-
9
914
BX647985.1
-
9
943
CR748189.1
-
6
348
DA156504.1
-
4
227
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0