Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGIF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199513.1 | NP_001186442.1 | 3 | 748 | ||
NM_001199514.1 | NP_001186443.1 | 3 | 876 | |||
NM_001199515.1 | NP_001186444.1 | 3 | 778 | |||
NM_021809.6 | NP_068581.1 | 3 | 835 | |||
GenBank mRNA | AB042646.1 | - | 3 | 844 | ||
AK074802.1 | - | 3 | 815 | |||
AK074829.1 | - | 1 | 330 | |||
AK313919.1 | - | 3 | 835 | |||
BC006549.2 | - | 3 | 822 | 2964507 | ||
BC012816.2 | - | 3 | 822 | 2964507 |