Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021818.3 | NP_068590.1 | 2 | 584 | ||
XM_011537057.2 | XP_011535359.1 | 3 | 647 | |||
XM_017021568.1 | XP_016877057.1 | 3 | 877 | |||
GenBank mRNA | AJ292969.1 | - | 2 | 363 | ||
AK021500.1 | - | 2 | 531 | |||
AK023071.1 | - | 2 | 413 | |||
AK095903.1 | - | 3 | 448 | |||
AK225896.1 | - | 2 | 556 | |||
AK290883.1 | - | 3 | 456 | |||
BC020537.1 | - | 2 | 530 | 4425620 | ||
CR457297.1 | - | 2 | 198 |