Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTKD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021826.4 | NP_068598.1 | 2 | 665 | ||
GenBank mRNA | AB058695.1 | - | 1 | 3031 | ||
AK023211.1 | - | 2 | 644 | |||
AK301748.1 | - | 2 | 617 | |||
BC007413.2 | - | 2 | 665 | 3010170 | ||
BC011856.1 | - | 2 | 629 | 4298806 |