Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NYX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029709.1 | XP_016885198.1 | 3 | 2313 | ||
GenBank mRNA | AJ278865.1 | - | 2 | 2671 |