Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021926.3 | NP_068745.2 | 2 | 642 | ||
GenBank mRNA | AB058691.1 | - | 2 | 545 | ||
AF294629.1 | - | 2 | 642 | |||
AJ404888.1 | - | 2 | 590 | |||
AK092346.1 | - | 2 | 667 |